Would you like to log in again?
pBC Phagemid Vectors, Agilent Technologies
Supplier: Agilent Technologies
212216 212216 212215 212218 212217 212218 212215 212217
99900-426EA 534.48 USD InStock
CA99900-424 99900-426 99900-428 CA99900-426 99900-424 CA99900-394 CA99900-428 99900-394
pBC Phagemid Vectors, Agilent Technologies
Vectors, Plasmids and Libraries
The pBC vectors were derived from the pBluescript II phagemid. The ampicillin-resistance gene has been replaced with the chloramphenicol resistance gene.
  • Convenient selection when subcloning from ampicillin-resistant cloning vectors
  • Large and versatile polylinker in two orientations for cloning flexibility
  • Available with f1 origin in either (+) or (–) orientation
pBC phagemids (plasmids with a phage origin) are cloning vectors designed to simplify commonly used cloning and sequencing procedures, including the construction of nested deletions for DNA sequencing, generation of RNA transcripts in vitro and site-specific mutagenesis and gene mapping.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty