Would you like to log in again?
StemBoost™ Reprogramming Cocktail, SMC4, Biovision
Supplier: BioVision
AB286242-5 AB286242-5 AB286242-1ML
76270-714EA 950.87 USD InStock
76270-714 76270-712 76270-710
StemBoost™ Reprogramming Cocktail, SMC4, Biovision
Cell Culture Supplements and Additives
SMC4 is a Small Molecule Cocktail of 4 inhibitors in DMSO solution. This media additive supports the derivation of hiPSCs (Human induced Pluripotent Stem Cells) in feeder or feeder-free culture environments. Use of this media additive increases efficiency of reprogramming, enhances clonality and tolerance to single cell and enables enzymatic passaging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty