Would you like to log in again?
StemBoost™ Fate Maintenance Medium (FMM) Cocktail, Biovision, Inc.
Supplier: BioVision
AB286243-5ML AB286243-5ML AB286243-1ML
76270-718EA 1493.45 USD InStock
76270-718 76270-716 76270-720
StemBoost™ Fate Maintenance Medium (FMM) Cocktail, Biovision, Inc.
Cell Culture Supplements and Additives
FMM cocktail is a cocktail of three small molecules in DMSO. This media additive along with basic fibroblast growth factor (bFGF) and leukemia inhibitory factor (LIF), supports the continuous footprint-free hiPSCs passaged as single cells in a feeder-free environment without the additional requirement of transgene expression.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty