Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AS-703026

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: 0.0 out of 5

Sort all by:

Synonyms: (S)-N-(2,3-Dihydroxypropyl)-3-((2-fluoro-4-iodophenyl)amino)isonicotinamideMSC1936369BPimasertib

AB235569-25MG AB235569-5MG
76268-468EA 543.94 USD InStock
76268-468 76268-466
AS-703026
AS-703026
Pimasertib, also known as AS703026, SAR245509, MSC1936369B, is a n orally bioavailable small-molecule inhibitor of MEK1 and MEK2 (MEK1/2) with potential antineoplastic activity.


Formula: C₁₅H₁₅FIN₃O₃
MW: 431.2 g/mol
MDL Number: MFCD16660676
CAS Number: 1236699-92-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/81/24105381.jpg
Order Now

 

ORDER

DOCUMENTATION

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty