Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

LY2228820 ≥98%

Supplier: BioVision
Synonyms: 5-(2-(tert-Butyl)-4-(4-fluorophenyl)-1H-imidazol-5-yl)-3-neopentyl-3H-imidazo[4,5-b]pyridin-2-amine dimethanesulphonate5-(2-(tert-Butyl)-4-(4-fluorophenyl)-1H-imidazol-5-yl)-3-neopentyl-3H-imidazo[4,5-b]pyridin-2-amine dimethanesulfonateRalimetinib dimesylateLY2228820 dimesylate5-[2-tert-butyl-4-(4-fluorophenyl)-1H-imidazol-5-yl]-3-(2,2-dimethylpropyl)imidazo[4,5-b]pyridin-2-amine methanesulphonic acid5-[2-tert-butyl-4-(4-fluorophenyl)-1H-imidazol-5-yl]-3-(2,2-dimethylpropyl)imidazo[4,5-b]pyridin-2-amine methanesulfonic acid

AB143833-25MG AB143833-5MG
76268-778EA 271.39 USD InStock
76268-778 76268-780
LY2228820 ≥98%
LY2228820
LY2228820 is a novel and potent inhibitor of p38 MAPK with IC50 of 7 nM in a cell-free assay, but does not alter p38 MAPK activation.


Formula: C₂₆H₃₇FN₆O₆S₂
MW: 612.74 g/mol
MDL Number: MFCD28101032
CAS Number: 862507-23-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/85/24105385.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty