Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Kobe2602 ≥98%

Supplier: BioVision
Synonyms: 1-[2,6-dinitro-4-(trifluoromethyl)anilino]-3-(4-fluorophenyl)thioureaKobe2602Kobe-2602kobe-26022-(2,6-dinitro-4-(trifluoromethyl)phenyl)-N-(4-fluorophenyl)hydrazinecarbothioamideKobe 2602kobe 2602

AB235509-5MG AB235509-25MG
76268-498EA 743.11 USD InStock
76268-498 76268-496
Kobe2602 ≥98%
Kobe2602
Kobe2602 is a novel and effective small-molecule compound inhibiting Ras–Raf interaction by SBDD; exhibits potent activity to competitively inhibit the binding of H-Ras·GTP to c-Raf-1 RBD with a Ki value of 149 ± 55 μM.


Formula: C₁₄H₉F₄N₅O₄S
MW: 419,31 g/mol
CAS Number: 454453-49-7
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/93/24105393.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty