Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

AZ 628 ≥98%

Danger
Synonyms: 3-(2-cyanopropan-2-yl)-N-[4-methyl-3-[(3-methyl-4-oxoquinazolin-6-yl)amino]phenyl]benzamideAZ 628AZ-628AZ628

AB235573-5MG AB235573-25MG
76268-510EA 1126.14 USD InStock
76268-510 76268-508
AZ 628 ≥98%
AZ 628
AZ628 is a new pan-Raf inhibitor for BRAF, BRAFV600E, and c-Raf-1 with IC50 of 105 nM, 34 nM and 29 nM, also inhibits VEGFR2, DDR2, Lyn, Flt1, FMS, etc.


Formula: C₂₇H₂₅N₅O₂
MW: 451,53 g/mol
MDL Number: MFCD17392577
CAS Number: 878739-06-1
https://us-legacy.vwr.com/stibo/web/std.lang.all/53/96/24105396.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty