Would you like to log in again?
MBS Mammalian Transfection Kit, Agilent Technologies
Supplier: Agilent Technologies
200388 200388
ST200388EA 607.21 USD InStock
CAST200388 ST200388
MBS Mammalian Transfection Kit, Agilent Technologies
Transfection Reagents
The MBS Mammalian Transfection kit can increase transfection efficiencies up to 100-fold.
Cells are incubated with CaPO4/DNA coprecipitate for only 3 hours, versus 6 to 12 hours with traditional CaPO4 methods.
Additionally, cells can be assayed 12 to 24 hours post-transfection instead of the usual 48 to 72 hours.
For highest viral titer use this kit to transfect HEK293 cells. Solution 1: 2.5 M CaCl2, Solution 2: 2x BBS (pH 6.95), Solution 3: Modified bovine serum (MBS), Control Plasmid: pCMV, Beta-gal.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty