Would you like to log in again?
T4 DNA Ligase, Agilent Technologies
Supplier: Agilent Technologies
600011 600011
ST600011EA 249.35 USD InStock
ST600011 CAST600011
T4 DNA Ligase, Agilent Technologies
Nucleic Acid Reagents Nucleic Acid Modification and Labelling
T4 DNA ligase is used to ligate cohesive ends in an overnight reaction.
  • High efficiency at joining fragments with complementary sticky ends
It catalyzes the linkage of adjacent 5'-phosphate and 3'-hydroxy ends of double-stranded DNA by formation of a phosphodiester bond.
Caution: For research use only. Not for use in diagnostic procedures.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty