Would you like to log in again?
VWR® Standard Manual Defrost Chest Freezers
Supplier: VWR
VWR®
MFV-15-C MFV-20-C
76285-198EA 3070.29 USD InStock
76285-198 76285-196
VWR® Standard Manual Defrost Chest Freezers
Freezers Laboratory / General Purpose Freezers
VWR® Standard Series Manual Defrost Laboratory Chest Freezers are designed specifically with general-purpose applications in mind.
  • LED interior Light
  • Keyed door lock
  • Magnetic door gasket
These units are ideal when stable temperatures averaging −20 °C are required but precise temperature control, maintenance, and alarms are not necessary. These freezers feature a manual defrost design to eliminate damaging temperature spikes from defrost cycles. This ensures valuable sample security. Defrosting of the unit must be performed periodically to prevent severe ice build-up.
Units feature keyed door lock, LED interior lights and high-density urethane foam cabinet and door insulation.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
SERVICES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty