Would you like to log in again?
Centaur Single-Channel Pipettors, Labsciences
Supplier: Labsciences
5000-VV 10000-FV
11020-002EA 187.19 USD InStock
11020-002 11020-004
Centaur Single-Channel Pipettors, Labsciences
Pipettes Micropipettes
Centaur pipettors feature quick, easy volume setting and lightweight design with a soft plunger movement to minimize hand fatigue and RSI.
  • Click stop digital adjustable volume mechanism
  • Streamlined tip ejector for easy access to narrow containers/tubes
  • Easy to calibrate and service
  • Three year warranty on all components
Two stage plunger operation for 'blow-out' step during pipeting. It uses Industry Standard pipette tips.
Certifications: Individually calibrated in accordance with DIN 12650 Standards.
Ordering information: Special volume ranges are available on request.
Delivery information: Supplied with a calibration tool and computer generated calibration report.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty