Would you like to log in again?
Universal Flex SideShields™, Eyewears, Radians
Supplier: Radians
FLEX SIDESHIELD™
99705 99706
89218-352PK 589.77 USD InStock
89218-352 89217-628
Universal Flex SideShields™, Eyewears, Radians
Safety Glasses
These Sideshields are a quick way of supplying additional protection to visitors and employees wearing personal priscription eyewear.
  • Easy on and off, weaves through the temple
  • Blocks 100% of UV
  • Molded to a bendable softness
Engineered to fit all types of eyewear by easily attaching to temples and fronts. No parts or tool required. Absorbs the energy of impact and allows temples to fold correctly.
Certifications: Conforms to ANSI Z87.1 Standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty