Would you like to log in again?
96 microTUBE Plate, Covaris
Supplier: COVARIS INC MS
520078
MSPP-520078EA 961.54 USD InStock
MSPP-520078
96 microTUBE Plate, Covaris
Microplates Deep Well Microplates
96 microTUBE plate is designed for the automated batch processing capability of the Ultrasonicators.
  • Made of a special borosilicate glass for low impedance and better transmission of acoustic energy
  • No cross contamination, clean-up, or aerosol formation
  • Supplied in single use pre-pierced plate
  • Pre-assembled, ready to use
  • Delivers sheared DNA of the optimal size for the sequencer, by unbiased, and non-damaging
  • No heat damage, so sample quality and labile metabolites are preserved
The microTUBE is widely used to shear DNA and RNA into fragments with distributions centered from 100 to 1,500 bp. The microTUBE is also ideally suited for other applications with AFA in small volumes such as chromatin shearing for ChIP, tissue disruption and homogenization for biomarker discovery and ADME/Tox screening.
The integrated AFA fiber helps nucleation of cavitation bubbles and allows more uniform sample processing, which in turn increases the efficiency and reproducibility of nucleic acid shearing. The result is the simultaneous collapse of millions of high energy bubbles that allow microTUBE to accurately, precisely, and controllably exert mechanical energy on sample. This energy is able to shear DNA and RNA down to 100 bp one magnitude faster than other technologies. Focused-ultrasonicators utilizing AFA technology is also used extensively to shear chromatin and disrupt tissues.
Certifications: Meets SBS (Society of Biomolecular Screening specifications).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty