Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
'Ivy' Trainer Pad
Supplier: WALLCUR, LLC.
Ivy
REM-IV-PAD-01L
470306-778EA 414.19 USD InStock
470306-778
'Ivy' Trainer Pad
Models Human Biology Models
Realistic Blood Return!
Remedy's simulated IV pad was designed by Janet McGuigan, BSN, RN, CHSE. "Ivy" can be used for the training and practice of IV fluid and medication administration and flushing. Any standardized patient, task trainer or mannequin can be used with no special set up or clean up. There is realistic blood return and no special fluids are required. "Ivy" can be used with any IV pump and tubing and is an economical, efficient way to practice setting IV flow rates. Catheter in place. Not intended for IV insertion simulation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty