Would you like to log in again?
Calbiochem® Streptavidin
Supplier: EMD Millipore
Calbiochem®
189730-1MG 189730-10MG
EM189730-1MGEA 181.49 USD InStock
EM189730-1MG EM189730-10MG
Calbiochem® Streptavidin
Proteins and Peptides
  • A high-affinity biotin binding protein
  • Unit of definition: One unit is defined as the amount of streptavidin required to bind 1.0 µg of D-Biotin at pH 7.0
  • Specific activity: ≥10 units/mg protein
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty