Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cd8a Rabbit Monoclonal Antibody [clone: YTS 105.18]

Supplier: Enzo Life Sciences
ABS456-0200
76284-780EA 600.38 USD InStock
76284-780
Anti-Cd8a Rabbit Monoclonal Antibody [clone: YTS 105.18]
Antibodies
Recombinant monclonal antibody (Rabbit IgGκ).

Recommended Applications: Flow Cytometry, IF, IHC (FS), FUNC

Type: Primary
Antigen: Cd8a
Clonality: Monoclonal
Clone: YTS 105.18
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGκ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty