Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Cd8b Rat Monoclonal Antibody [clone: YTS 156.7.7]

Supplier: Enzo Life Sciences
ABS631-0200
76285-122EA 622 USD InStock
76285-122
Anti-Cd8b Rat Monoclonal Antibody [clone: YTS 156.7.7]
Antibodies
Recombinant monclonal antibody (Rat IgG2bκ).

Recommended Applications: Flow Cytometry, IHC

Type: Primary
Antigen: Cd8b
Clonality: Monoclonal
Clone: YTS 156.7.7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2bκ
Reactivity: Mouse

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty