Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MUC1 Rabbit Monoclonal Antibody [clone: HMFG2]

Supplier: Enzo Life Sciences
ABS644-0200
76285-148EA 585.76 USD InStock
76285-148
Anti-MUC1 Rabbit Monoclonal Antibody [clone: HMFG2]
Antibodies
Recombinant monclonal antibody (Rabbit IgGλ).

Recommended Applications: ELISA, IHC

Type: Primary
Antigen: MUC1
Clonality: Monoclonal
Clone: HMFG2
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgGλ
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty