Would you like to log in again?
SureLINK™ AP Conjugation Kit, Seracare
Supplier: SeraCare
5610-0026 5610-0025
95059-508EA 1521.95 USD InStock
95059-508 95059-506
SureLINK™ AP Conjugation Kit, Seracare
Assays Immunological Assays
KPL SureLINK AP Conjugation kits enables antibody or protein labeling with alkaline phosphatase (AP) in only three hours, saving several hours over standard maleimide/thiol methods.
  • Stored at 2 - 8 °C
  • Stable for one year when stored as recommended
Sufficient reagents are provided to conjugate three samples, each containing between 0.05 to 0.1 mg or 0.1 to 0.5 mg of antibody or protein.
Delivery information: Kit contains all reagents required to conjugate a protein or antibody, including ready-to-use buffers and an optimized storage buffer. Modified AP is supplied ready-to-use and requires no activation or purification.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty