Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aluminium oxide ≥97%, GREENCAST® 97 PLUS

Supplier: HARBISON-WALKER
Synonyms: Aluminium(III) oxideAlumina C

59338
80700-882EA 483.3 USD InStock
80700-882
Aluminium oxide ≥97%, GREENCAST® 97 PLUS
Aluminium oxide
GREENCAST® 97 PLUS is a 97% alumina dense castable for temperatures up to 3400 °F (1870 °C). It is based on very high purity ingredients, low in silica, iron oxide and alkalies.


Formula: Al₂O₃
MW: 101.96 g/mol
MDL Number: MFCD00003424
CAS Number: 1344-28-1
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty