Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Airfree® Schlenk Connecting Adapters, Inlet, 90° Angle, Chemglass

Supplier: Chemglass
Airfree®
AF-0072-01
80064-260EA 227.88 USD InStock
80064-260
Airfree® Schlenk Connecting Adapters, Inlet, 90° Angle, Chemglass
Adapters Adapters, Joints with Tube Connectors
Connecting inlet adapter.

  • Equipped with a #15 O-Ring joint
  • Equipped with a 0 to 4 mm, 1-arm Chem-Vac™ Chem-Cap® valve

Side arm is 9 mm O.D. medium-wall tubing for use with 3/8" Cajon® connections. Approximate overall height is 100 mm.

Delivery information: Each adapter is supplied complete with one #116 Viton O-Ring.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty