Would you like to log in again?
Airfree® Schlenk Vacuum Trap, Chemglass
Supplier: Chemglass
Airfree®
AF-0300-20
80064-412EA 631.9 USD InStock
80064-412
Airfree® Schlenk Vacuum Trap, Chemglass
Vacuum Traps
Two-piece vacuum trap with # 20 O-Ring joints on both the inlet and outlet ports.
  • Trap body is 57 mm O.D.×300 mm in length
  • Arms on stopper portion connect to 80064-398 manifold and 80064-402 shut off adapters
The seal between the trap stopper and body is made with a 57 mm flat O-Ring flange for total grease-free operation. Used with a 4300 ml dewar flask (80061-498 or 80061-508).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty