Would you like to log in again?
Premiere Slide Warmers for Histology, Newcomer Supply
Supplier: Newcomer Supply
Premiere
6259 6262 6261 6258
76278-844EA 465.78 USD InStock
76278-844 76278-804 76278-846 76278-802
Premiere Slide Warmers for Histology, Newcomer Supply
Slide Warmers/Dryers
Ideal for use in histology, cytology, pathology and biology for drying and mounting paraffin tissue sections on slides.
  • Temperature range room temperature to 75 °C
  • Anodized black surface provides contrast
  • LED temperature display
  • Available in 2 sizes with or without lid
These slide warmers are easy to adjust and use. The thermal heater ensures even heat transfer.
Certifications: The slide warmers are CE certified and the electrical cord is CSA and UL approved.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty