Would you like to log in again?
Cardiolipin probe, Biovision, Inc.
Supplier: BioVision
AB285385-5MG AB285385-25MG
76269-158EA 306.43 USD InStock
76269-158 76269-160
Cardiolipin probe, Biovision, Inc.
Protein Linking and Labeling Systems
Cardiolipin probe is an excellent fluorescent probe for the detection and quantification of cardiolipin (CL). It displays improved sensitivity and excellent selectivity for CL over other major mitochondrial membrane lipids.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty