Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-Bsn Mouse Monoclonal Antibody [clone: SAP7F407]

Supplier: Enzo Life Sciences
ABS666-0200 ABS666-0050
76271-374EA 451.39 USD InStock
76271-374 76271-376
Anti-Bsn Mouse Monoclonal Antibody [clone: SAP7F407]
Antibodies
Recommended Applications: ICC, IF, IHC, IP, WB

Type: Primary
Antigen: Bsn
Clonality: Monoclonal
Clone: SAP7F407
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2aκ
Reactivity: Human;Mouse;Rat;Dog;Bovine

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty