Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-MT1A Mouse Monoclonal Antibody [clone: UC1MT]

Supplier: Enzo Life Sciences
ABS681-0200
76288-364EA 751.46 USD InStock
76288-364
Anti-MT1A Mouse Monoclonal Antibody [clone: UC1MT]
Antibodies
Recommended Applications: ELISA, IHC (PS), WB

Type: Primary
Antigen: MT1A
Clonality: Monoclonal
Clone: UC1MT
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat;Rabbit;Horse;Fish

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty