Would you like to log in again?
Magid® ROC® Nitrile Palm Coated Gloves, Magid
Supplier: Magid
GP16010 GP1608 GP1609 GP16011 GP1607
69100-301PK 48.66 USD InStock
69100-301 69100-303 69100-299 69100-305 69100-307
Magid® ROC® Nitrile Palm Coated Gloves, Magid
Gloves
Magid® ROC® work gloves are a great alternative to heavier cotton and canvas coated gloves. The GP160 features a grey nitrile palm coating to ensure a tight grip and resists cuts and abrasions.
  • Scratch-resistant grey nitrile palm coating
  • Nylon knit wrist cuff
  • Excellent for general maintenance duties
This soft glove also features a white 100% polyester machine knit shell and knitted wrist cuff, which provides flexibility and breathability for a comfortable fit.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty