Would you like to log in again?
Magid® ROC® Polyurethane Palm Coated Gloves, Magid
Supplier: Magid
JPS28 JPS211 JPS29 JPS27 JPS210
69100-457PK 45.47 USD InStock
69100-457 69100-465 69100-463 69100-461 69100-459
Magid® ROC® Polyurethane Palm Coated Gloves, Magid
Gloves
Work gloves are flexible, form fitting polyurethane coated gloves that feel like you're working with bare hands. The gloves feature a seamless machine knit, white 100% polyester shell and knit wrist cuff.
  • Breathable, 100% polyester shell
  • Abrasion-resistant polyurethane palm coating
  • Designed for lint free applications
  • ANSI Cut Level 1 and ANSI Abrasion 2
The polyurethane palm coating provides excellent grip and dexterity.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty