Would you like to log in again?
Magid® ComfortFlex® Smooth Finish Nitrile Gloves, Magid
Supplier: Magid
Comfort Flex®
NB21XXL NB21XL NB21L
69100-455PK 108.57 USD InStock
69100-455 69100-453 69100-451
Magid® ComfortFlex® Smooth Finish Nitrile Gloves, Magid
Gloves
The Magid ComfortFlex is one of the most durable work gloves available. With their knit-lined nitrile construction, ComfortFlex gloves provide workers barrier protection from chemicals, oil and grease as well as abrasion resistance.
  • Nitrile coating offers excellent chemical resistance
  • Textured palm for premium grip in wet and oily conditions
  • Comfortable, seamless knit liner
  • 12" long for protection well past the wrist
The fully washable glove comes in a variety of hand sizes for nearly any general-purpose industrial setting or application. With its premium style, functionality and comfort, ComfortFlex gloves are as versatile as they are durable and protective.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty