Would you like to log in again?
Magid® SparkGuard® Cape Sleeve, Magid Glove
Supplier: Magid
SparkGuard®
1855S 1855L 1855M 1855XL
69100-059EA 28.7 USD InStock
69100-059 69100-057 69100-061 69100-063
Magid® SparkGuard® Cape Sleeve, Magid Glove
Workwear
Magid SparkGuard Flame Resistant Cape Sleeve is made from fabric that self extinguishes when the ignition source or flame has been removed.
  • Collar
  • Adjustable wrist snaps
Made with 100% cotton sateen with six snaps so it can be attached to Magid bib style (69100-069).
This sleeve offers a meaningful degree of flame resistant protection at a reasonable cost while comfortable to wear while working.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty