Would you like to log in again?
Magid® TerryMaster Loops-Out Heavyweight Terrycloth Glove, Magid
Supplier: Magid
PT930R
11000-246EA 10.88 USD InStock
11000-246
Magid® TerryMaster Loops-Out Heavyweight Terrycloth Glove, Magid
Gloves
Magid® TerryMaster loops-out heavyweight terrycloth gloves are seamless, machine knit gloves designed for hard core protection.
  • Economical alternative to hot mill gloves
  • Extra-heavyweight terrycloth glove
  • Excellent heat and thermal protection
This glove features a 30 oz. extra heavyweight blend of cotton and polyester machine-knitted into sturdy, loops out terrycloth. Terrycloth provides the exceptional cut, abrasion, and thermal protection.
Professional testing rates this glove at ANSI cut level 2, giving the PT930R a similar level of protection to aramid and hppe fibers, but for a fraction of the cost.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty