Would you like to log in again?
Magid® TouchMaster® 650H/651H Lightweight Cotton Lisle Inspection Glove, Magid
Supplier: Magid
650H 651H
11000-252CS 1501.32 USD InStock
11000-252 11000-296
Magid® TouchMaster® 650H/651H Lightweight Cotton Lisle Inspection Glove, Magid
Gloves
The inspection glove is 100% cotton two-piece lisle glove constructed of cool and breathable cotton.
  • Two-piece cotton construction
  • Cut and sewn pattern
  • Color-coded sizes for easy sorting
  • Hemmed edge to prevent unraveling
This glove is form fitting and it features a hemmed edge to prevent unraveling in the wash. The glove can be worn alone to protect delicate products from smudges, finger prints or skin oils, or it can be used as a liner to absorb moisture. This glove is a great general-purpose glove for light jobs in any industry.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty