Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

7-Hydroxy-4-methylcoumarin-3-acetic acid, succinimidyl ester

Supplier: Anaspec Inc
Synonyms: N-Succinimidyl 7-Hydroxy-4-methylcoumarin-3-acetate7-Hydroxy-4-methylcoumarin-3-acetic acid N-Succinimidyl ester4-Methylumbelliferone-3-acetic acid N-Succinimidyl esterN-Succinimidyl 4-Methylumbelliferone-3-acetate

AS-81239
103010-924EA 220.69 USD InStock
103010-924
7-Hydroxy-4-methylcoumarin-3-acetic acid, succinimidyl ester
7-Hydroxy-4-methylcoumarin-3-acetic acid succinimidyl ester
7-Hydroxy-4-methylcoumarin-3-acetic acid, SE is a blue fluorophore that has pH-dependent and environment-sensitive fluorescence. It is widely used for preparing bioconjugates of blue fluorescence.

  • Spectral properties: Abs/Em = 364/458 nm
  • Solvent system: DMF or DMSO
  • Storage Temperature: -20°C desiccated and protected from light
  • Shipping condition: Blue ice


Formula: C₁₆H₁₃NO₇
MW: 331.28 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00467590
CAS Number: 96735-88-5
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty