Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Anti-IgG Fc Mouse Monoclonal Antibody (Biotin) [clone: N3318]

Supplier: TCI
M3053-VIAL
TCM3053-VIALEA 339 USD InStock
TCM3053-VIAL
Anti-IgG Fc Mouse Monoclonal Antibody (Biotin) [clone: N3318]
Antibodies
The monoclonal antibodies recognize the Fc region of human IgG.

This antibody reacts with human IgG on ELISA (1:5000) and Western blotting (1:1000).

Type: Primary
Antigen: IgG Fc
Clonality: Monoclonal
Clone: N3318
Conjugation: Biotin
Epitope: F(c)
Host: Mouse
Isotype: IgG1
Reactivity: Human

Order Now
 

ORDER

SPECIFICATIONS

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty