Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Human recombinant TBK1 (from Baculovirus (Insect cells))
Supplier: Cayman Chemical
22817-50
76292-900EA 799.17 USD InStock
76292-900
Human recombinant TBK1 (from Baculovirus (Insect cells))
Enzymes
TANK-binding kinase 1 (TBK1) is a noncanonical inhibitor of NF-κB kinase (IKK) that has an essential role in regulating inflammatory responses to pathogens. Following activation of toll-like receptors by viral DNA, TBK1 interacts with various partners such as STING, MAVS, and TANK to phosphorylate and activate interferon regulatory factors (IRFs) 3 and 7 as well as DEAD-box helicase 3 X-linked (DDX3X), which leads to transcriptional activation of pro-inflammatory and antiviral genes including interferon (IFN) subtypes α and β. TBK1 induces nuclear translocation of NF-κB to initiate a pro-inflammatory response <em>via</em> phosphorylation of NF-κB inhibitor α (NFκBIα), IKK2, or NF-κB p65 subunit (RELA). Cytosolic localization of <em>E. coli</em>, <em>Salmonella</em>, and <em>S. pyogenes</em> increases in TBK1<sup>-/-</sup> murine embryonic fibroblasts, macrophages, and epithelial cells, suggesting TBK1 maintains vacuolar integrity that is critical to bacterial clearance. TBK1 also phosphorylates the autophagy receptor optineurin to enhance binding of ubiquitin-like microtubule-associated protein light chain 3 (LC3) modifiers and induce autophagic clearance of <em>S. enterica</em>.
Used for: Antivirals, Autoimmunity, Innate Immunity, NF-kB
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty