Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
S396A mutant (from E. coli)
Supplier: Cayman Chemical
23590-100
76292-904EA 578.72 USD InStock
76292-904
S396A mutant (from E. coli)
Enzymes
IFR3 (S386A, S396A mutant; human recombinant) contains amino acids corresponding to human IRF3 (Item No. 22811) with alanine substituted for serine at positions 386 and 396. Interferon regulatory factor 3 (IRF3) is a member of the IRF family that plays a crucial role in activation of innate immunity and inflammation in response to viral infection, functioning as a molecular switch for antiviral activity. Double-stranded RNA generated during a viral infection leads to IRF3 activation through serine/threonine phosphorylation by TBK1 (Item No. 22817) or IKKε (IKBKE) kinases, which induces a conformational change leading to its dimerization, nuclear localization, and association with CREB binding protein (CREBBP)/p300. The complex formed by this association, known as DRAF1, activates transcription of interferon α (IFN-α) and IFN-β as well as other IFN-induced genes, which play a critical role in the type 1 IFN-dependent immune response. Various serine residues have been implicated in IRF3 activation, including S386 and S396. TBK1 and IKKε phosphorylation of IRF3 is decreased when serine is replaced with alanine at positions 396, 398, 402, and 405 and with threonine at position 404. Phosphorylation of S386 is essential for IRF3 oligomerization and binding to p300, and phosphorylation of S396, T390, and either S385 or S386 occurs following Sendai viral infection in HEK293 cells.
Used for: Autoimmunity, Innate Immunity
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty