Would you like to log in again?
Magid® Kevlar String Knit Gloves, Magid
Supplier: Magid
13651KEVKW
69100-104CS 1883.8 USD InStock
69100-104
Magid® Kevlar String Knit Gloves, Magid
Gloves
Kevlar string knit gloves offer great cut resistance and heat protection.
  • Overcast band keeps glove from unraveling and offers durability
  • Color coded overcast band allows for easy pairing when washed
  • Gloves offer great hand protection for everyday use
These gloves are ambidextrous, allowing the ability to interchange the gloves and evenly distribute the use and abuse on glove endures. This feature ensures that the gloves last longer. Also enhancing the gloves' longevity is the white overcast band which prevents the glove from unraveling.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty