Would you like to log in again?
Syringe Filters, Filtropur S 0.2, Sarstedt
Supplier: Sarstedt
83.1826.001
103573-246PK 132.52 USD InStock
103573-246
Syringe Filters, Filtropur S 0.2, Sarstedt
Filters Syringe Filters
Filtropur S 0.2 syringe filter with PES membrane, without pre-filter.
  • Gamma irradiated
  • Non-pyrogenic, endotoxin-free, non-cytotoxic
Ideal for sterile filtration.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty