Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Platinum replacement target (for EMS 650, 675X)

Supplier: Electron Microscopy Sciences
Synonyms: Platinium

91032
102091-406EA 4175.41 USD InStock
102091-406
Platinum replacement target (for EMS 650, 675X)
Platinum
ELECTRON MICROSCOPY SCIENCES Replacement Targets

Recommended change as set of three


Formula: Pt
MW: 195.08 g/mol
MDL Number: MFCD00011179
CAS Number: 7440-06-4
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty