Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Trichloroacetic acid (TCA) 30% (w/v)

Supplier: Ricca Chemical
Danger
Synonyms: TCAAceto-CaustinTrichloroethanoic acid

R8688000-500C
RCR8688000500CEA 168.53 USD InStock
RCR8688000500C
Trichloroacetic acid (TCA) 30% (w/v)
Trichloroacetic acid (TCA)


Formula: Cl₃CCOOH
MW: 163.39 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004177
CAS Number: 76-03-9
UN: 2564
ADR: 8,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/74/52/727452.jpg
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. Sodium Hydroxide/Phenolphthalein) 29.6-30.4 % (w/v)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty