Would you like to log in again?
Proto® Torqueplus™ 6-Point Short Combination Wrenches, Essendant
Supplier: Stanley
577-1205EFS
76188-602EA 24.78 USD InStock
76188-602
Proto® Torqueplus™ 6-Point Short Combination Wrenches, Essendant
Wrenches
Applies more torque to fastener.
  • Torqueplus™ fastening system drives off fastener body, more easily removing damaged fasteners
  • Reduces risk of fastener rounding
  • Unique arc design increases surface contact
  • Anti-Slip Design (ASD) wrench has a locking groove to prevent slippage and knuckle injury
  • Shank is positioned at the center of box end; reduces the tendency of the wrench to cock
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty