Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Tech-Med® Exam Lamp, DUKAL Corporation®

Supplier: Dukal
4398 4399 4398M
76235-490EA 289.01 USD InStock
76235-490 76235-362 76235-360
Tech-Med® Exam Lamp, DUKAL Corporation®
Lights Examination Lights
12" flexible gooseneck lamps with chrome plated finish.

  • Deluxe and Mobile versions include wire guard and heat shield to keep lamp cool
  • 7' power cord included
  • Stationary lamps have a base diameter of 11"
  • Locking height adjustment on all lamps from 34 to 72"

Maximum wattage for all models: 60 watt incandescent bulb; 10 watt LED bulb; 13 watt SBCFL bulb.

Order Now
 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty