Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Tech-Med® Fracture Kit and Components, DUKAL Corporation®
Supplier: Dukal
Tech-Med®
8014 8016 8000 8012 8011 8015
76236-502EA 104 USD InStock
76236-502 76236-504 76236-506 76236-508 76236-510 76236-500
Tech-Med® Fracture Kit and Components, DUKAL Corporation®
First Aid Supplies First Aid Kits
In-line fiberglass support for transport and stabilization.
  • Splints are x-ray transparent
  • Limb splints can be used on patient's left or right side
  • Heavy-duty waterproof vinyl carry bag with storage compartments
Ordering information: Individual components are also available separately.
Delivery information: Complete Fracture Kit contains arm splint (adult and child), leg splint, (adult and child), and arm/shoulder immobilizer.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty