Would you like to log in again?
VWR® OTG Safety Glasses
Supplier: VWR
VWR®
OTG-R
76210-732CS 1637.11 USD InStock
76210-732
VWR® OTG Safety Glasses
Safety Glasses
Economical and compact, designed to fit over prescription glasses.
  • Fits over most prescription glasses
  • Adjustable temple length for varied facial contours
  • Polycarbonate lens
  • Provides 99.9% UV protection
The pantoscopic adjustment allows lens angle modification and it is impact-resistant.
Certifications: Meets ANSI Z87.1 standards.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty