Would you like to log in again?
Virkon®S Disinfectant Powder, Pharmacal
Supplier: PHARMACAL RESEARCH LABS MS
03010F
MSPP-03010FEA 124.77 USD InStock
MSPP-03010F
Virkon®S Disinfectant Powder, Pharmacal
Cleaning Agents Disinfectants
Virkon®S is a broad-spectrum disinfectant/cleaner for use in an animal research facility. It is fragrance-free and is available in easy-to-use packets of a very quick dissolving granular powder.
  • Used extensively as a vehicular undercarriage disinfectant/cleaner
  • Solutions are stable for 7 days after mixing
  • Virkon®S used at 1% will kill Canine, Feline, Porcine parvoviruses and FMD
  • Virkon®S used at 2% will kill Aspergillus
Effective against numerous microorganisms affecting animals, including viruses, gram positive and gram-negative bacteria, fungi (molds and yeasts), and Mycoplasma.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty