Would you like to log in again?
Pipette Can, Stainless Steel, Bellco Glass
Supplier: Bellco Glass
1217-22216
102681-544EA 88.58 USD InStock
102681-544
Pipette Can, Stainless Steel, Bellco Glass
Boxes Pipette Sterilization Containers
Pipette cans are constructed of heavy gauge stainless steel and are rectangular for easy stacking and storage of pipettes.
  • May be sterilized by autoclave or dry heat (do not exceed 400 °F).
Permanently bonded silicone pad on inside of top and bottom protects pipette tips and mouthpieces from chipping.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty