Would you like to log in again?
Bizflame 88/12 Flame Resistant Coveralls, Portwest
Supplier: Portwest
Bizflame™
UFR88NARXXL UFR88NARM UFR88NARL UFR88NARXXXL UFR88NARXL
76105-882EA 124.56 USD InStock
76105-882 76105-884 76105-880 76105-886 76105-878
Bizflame 88/12 Flame Resistant Coveralls, Portwest
Workwear
Guaranteed flame resistant for the life of the garment.
  • Dual hazard protection
  • Protects against radiant and convective heat
  • Full front, top quality brass zipper
  • Bizflame 88/12, 7 oz. fabric
These are designed with two front, two side, two back, one sleeve and one rule pocket.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty