Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Looped Soft Push Caps for Reaction Tubes, Sarstedt

Supplier: Sarstedt
65.109
101093-712PK 36.92 USD InStock
101093-712
Looped Soft Push Caps for Reaction Tubes, Sarstedt
Closures Test/Sample Tube Closures
Looped soft push cap for reaction tubes of 1.5 ml.

  • Suitable for micro tubes of 10.8 mm in diameter
  • Made of LDPE

Certifications: CE certified.

Packaging: 1000 pieces/bag.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty