Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
BBL™ Urease Broth, BD Life Sciences
Supplier: BD Life Sciences
BBL™
221098 221719 221719
90001-498PK 202.86 USD InStock
90001-498 CA90001-498 90001-232
BBL™ Urease Broth, BD Life Sciences
Media for Microbiology Prepared Media for Microbiology
Urease test broth is a differential medium for organisms, particularly members of the Enterobacteriaceae, on the basis of their ability to produce urease.
  • Peptone and dextrose provide nutrients
  • Reduced buffer content helps promote rapid growth
  • Urease base is also supplied as a filter sterilized 10X concentrated solution for use in preparing urease test broth in the user’s laboratory
Urease test broth can be used for the identification of bacteria on the basis of urea utilization and it is particularly recommended for the differentiation of members of the genus proteus from those of salmonella and shigella in the diagnosis of enteric infections.
When organisms utilize urea, ammonia is formed during incubation, which makes the reaction of these media alkaline, producing a pink-red color. Consequently, urease production may be detected by the change in the phenol red indicator.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty