Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Hydrochloric acid ≥32% (by titrimetric analysis), Puriss. p.a., Fluka™

Supplier: Honeywell Research Chemicals
Danger
Synonyms: Muriatic acid

84421-2.5L 84421-1L
BJ84421-2.5LEA 107.72 USD InStock
BJ84421-2.5L BJ84421-1L
Hydrochloric acid ≥32% (by titrimetric analysis), Puriss. p.a., Fluka™
Hydrochloric acid


Formula: HCl
MW: 36.46 g/mol
Boiling Pt: 42 °C (1.013 hPa)
Melting Pt: -35 °C
Density: 1.190 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011324
CAS Number: 7647-01-0
EINECS: 231-595-7
UN: 1789
ADR: 8,II
Order Now
Specification Test Results
Assay 32.0 - 35.0 %
Density (D 20/4) 1.150 - 1.170
Residue on ignition (SO₄) Max. 0.0005 %
Ammonium (NH₄) Max. 0.0001 %
Aluminium (Al) Max. 0.05 ppm
Arsenic (As) Max. 0.01 ppm
Barium (Ba) Max. 0.02 ppm
Beryllium (Be) Max. 0.02 ppm
Bismuth (Bi) Max. 0.1 ppm
Calcium (Ca) Max. 0.5 ppm
Cadmium (Cd) Max. 0.01 ppm
Cobalt (Co) Max. 0.01 ppm
Chromium (Cr) Max. 0.02 ppm
Copper (Cu) Max. 0.02 ppm
Iron (Fe) Max. 0.2 ppm
Germanium (Ge) Max. 0.05 ppm
Potassium (K) Max. 0.1 ppm
Lithium (Li) Max. 0.01 ppm
Magnesium (Mg) Max. 0.1 ppm
Manganese (Mn) Max. 0.01 ppm
Molybdenum (Mo) Max. 0.02 ppm
Sodium (Na) Max. 0.5 ppm
Nickel (Ni) Max. 0.02 ppm
Lead (Pb) Max. 0.02 ppm
Strontium (Sr) Max. 0.01 ppm
Titanium (Ti) Max. 0.1 ppm
Thallium (Tl) Max. 0.05 ppm
Vanadium (V) Max. 0.01 ppm
Zinc (Zn) Max. 0.05 ppm
Zirconium (Zr) Max. 0.1 ppm
Bromide (Br) Max. 50 ppm
Free chlorine (Cl) Max. 0.00005 %
Phosphate (PO₄) Max. 0.5 ppm
Sulfate (SO₄) Max. 1 ppm
Sulfite (SO₃) Max. 1 ppm
Vapor Pressure 3.23 psi ( 21.1 °C)

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty